Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH11 Recombinant Rabbit Monoclonal Antibody (17H3L51)

Catalog No. 702544
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-254-4 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-254-4 Supplier Invitrogen™ Supplier No. 702544
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Horse, Pig. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH11
Applications Western Blot
Classification Recombinant Monoclonal
Clone 17H3L51
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene MYH11
Gene Accession No. O08638, P35749, Q63862
Gene Alias AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain
Gene Symbols MYH11
Host Species Rabbit
Immunogen Peptides corresponding to human MYH11 [1) aa1915 -aa1933 2) aa1795 - aa1809].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17880, 24582, 4629
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.