Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564436 Supplier Invitrogen™ Supplier No. PA564436
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAGEQLKADL LLEPCSHYRF LTNGPSSSPG QERELFQETL ESLRVLGFSH EE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH14
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYH14
Gene Accession No. Q7Z406
Gene Alias 2400004E04Rik; DFNA4; DFNA4A; FP17425; II-C; KIAA2034; MHC16; Myh14; MYH14 variant protein; MYH17; myosin; myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy chain 14; myosin, heavy chain 14, non-muscle; myosin, heavy polypeptide 14; myosin-14; NHMCII; NMHC II-C; NMHC-II-C; nonmuscle myosin heavy chain; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; PNMHH
Gene Symbols MYH14
Host Species Rabbit
Immunogen Recombinant protein fragment of Human MYH14 (Product #RP-104174)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79784
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.