Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572848 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572848 Supplier Invitrogen™ Supplier No. PA572848
  • $594.00
    $478.65 / Each of 1

    Save $115.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibodies should not be exposed to prolonged high temperatures.

Muscle contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MYH3
Gene Accession No. P11055, P12847, P13541
Gene Alias DA2A; DA2B; DA8; embryonic skeletal myosin heavy chain isoform; HEMHC; Muscle embryonic myosin heavy chain; MYH3; MyHC-emb; Myhse; Myhs-e; MYHSE1; myosin heavy chain 3; Myosin heavy chain, fast skeletal muscle, embryonic; myosin heavy polypeptide 3; myosin, heavy chain 3, skeletal muscle, embryonic; myosin, heavy polypeptide 3, skeletal muscle, embryonic; myosin, skeletal, heavy chain, embryonic 1; myosin-3; RNMHCG; SMHCE
Gene Symbols MYH3
Host Species Rabbit
Immunogen MYH3 antibody for a 19 amino acid peptide near the amino terminus of human MYH3. The immunogen is within amino acids 30 - 80 of MYH3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17883, 24583, 4621
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.