Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ myImageAnalysis™ Software

Catalog No. PI62237
Change view
Click to view available options
Quantity:
2 licenses
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI62237 2 licenses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI62237 Supplier Thermo Scientific™ Supplier No. 62237

myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.

Thermo Scientific™ myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.

The myImageAnalysis Software comprises a complete set of easy-to-use research tools for the quantitative digital analysis of image data from electrophoresis gels and Western blots. The software uses a powerful algorithm to automatically select and identify lanes and band-boundaries for calculation of migration distances or densitometry. Intuitive, ready-to-use functions (buttons) provide molecular weight determination, relative and absolute quantitation, and purity calculation. Images are easily loaded, viewed, compared, adjusted (e.g., contrast, rotation, crop) and annotated. The program reads and writes to standard file formats, providing compatibility with nearly any source image or final presentation method.

Highlights:

  • Intuitive design sleek task-bars (ribbons) organize functions into four main tabs: File, Image, Lanes/Bands, Annotation

  • Compatible file types load and save images in popular file formats (TIFF, JPEG, PNG, etc.); analyze images from nearly any source, and easily share the results

  • Image comparison open two to six image files at once to review them simultaneously or toggle between them to edit them in parallel

  • Auto-identification accurate, customizable, automatic lane and band identification creates numbered, selectable objects for lane profile densitometry and analysis

  • MW determination versatile, trouble-free calculation of migration distances and molecular weights based on pre-installed or custom-added markers

  • Advanced quantitation automatically calculate sample purity based on band and lane intensities, or determine the relative or absolute quantities of bands, lanes or regions

  • Image refinement and annotation crop, straighten, invert and adjust (saturation, contrast, etc.); then add simple labels, notes and arrows

  • Exporting and sharing easily transfer data and images to Microsoft™ Excel™, Word™ and PowerPoint™ Programs for further analysis and presentation

Includes:

myImageAnalysis Software, 2 activation licenses, Quick Start Guide (within the software); user manual (within the software)

Requires:

Computer running MicroSoft™ Windows XP™, Windows Vista™, or Windows 7™ Operating System; not available for other operating systems at this time

Recommended for:

Automatically identify and delimit lane and band boundaries for analysis; Examine densitometry profiles of lanes and bands; Calculate and report migration distances and molecular weights of bands relative to standards; Overlay image of colored protein ladder onto image of chemiluminescent signal; Assess band purity by densitometry of stained bands relative to entire lanes; Measure and report the relative and absolute quantity of bands, lanes or regions; Quickly annotate and export analyzed images and quantitative data

TRUSTED_SUSTAINABILITY

Specifications

Product Type Software
Quantity 2 licenses
Content And Storage Contents: 1 software installation CD
For Use With (Equipment) myECL Imager
Product Line myImageAnalysis

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.