Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO18A Antibody [Alexa Fluor Plus 594], R&D Systems™
SDP

Catalog No. RD10016444
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10016444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10016444 Supplier R&D Systems Supplier No. NB10079840AFP594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

MYO18A Polyclonal antibody specifically detects MYO18A in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)

Specifications

Antigen MYO18A
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate Alexa Fluor Plus 594
Formulation 50mM Sodium Borate
Gene Alias KIAA0216DKFZp686L0243, MAJN, Molecule associated with JAK3 N-terminus, myosin 18A, Myosin containing a PDZ domain, myosin containing PDZ domain, myosin XVIIIA, myosin-XVIIIa, MYSPDZ, SP-A receptor subunit SP-R210 alphaS, SPR210
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 2004 andd 2054 of human myosin 18A using the numbering given in entry NP_510880.2 (GeneID 399687).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 399687
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.