Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577069 Supplier Thermo Scientific Supplier No. PA577069
Only null left
Add to Cart
Add to Cart

MYO1C Polyclonal Antibody

MYO1C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO1C
Gene Accession No. O00159, Q63355, Q9WTI7
Gene Alias C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic
Gene Symbols MYO1C
Host Species Rabbit
Immunogen Recombinant full length Human MYO1C.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17913, 4641, 65261
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.