Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565119 Supplier Invitrogen™ Supplier No. PA565119
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRVQRLQKKLE DQNKENHGLV EKLTSLAALR AGDVEKIQKL EAELEKAATH RRNYEEKGKR YRDAVEEKLA KLQKHNSELE TQKEQI Highest antigen sequence identity to the following orthologs - mouse 83%, rat 80%.

May be involved in transferrin trafficking. Likely to power actin-based membrane trafficking in many physiologically crucial tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO5C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO5C
Gene Accession No. Q9NQX4
Gene Alias 9130003O20Rik; BB085560; C330029I24; MYO5C; myosin 5C; myosin VC; myosin-Vc; unconventional myosin-Vc
Gene Symbols MYO5C
Host Species Rabbit
Immunogen Recombinant Human MYO5C. Recombinant protein control fragment (Product #RP-105882).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55930
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.