Learn More
Description
MA5-12902 targets MyoD1 in IF, IM, and IP applications and shows reactivity with Chicken, Human, mouse, and Rat samples. The MA5-12902 immunogen is recombinant mouse MyoD1 protein.
Specifications
Specifications
| Antigen | MYOD |
| Applications | Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5.8A |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | MYOD1 |
| Gene Accession No. | P10085, P15172, P16075, Q02346 |
| Gene Alias | AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
