Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myogenin Monoclonal Antibody (F5D), eBioscience™, Invitrogen™

Catalog No. p-7069448
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-131-11 25 μg
50-131-12 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-131-11 Supplier Invitrogen™ Supplier No. 14564380
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This F5D monoclonal antibody reacts with human, mouse, dog, and rat myogenin, a 34-kDa transcription factor. Expressed in skeletal and heart muscle, myogenin is a member of the MyoD family of basic-helix-loop-helix proteins, which also includes MyoD, Myf5, and MRF4. This transcription factor interacts with other helix-loop-helix proteins, which may or may not be muscle-specific. Myogenin plays a significant role in myogenic differentiation, even directing nonmuscle cells to the myogenic lineage. Transforming growth factor-beta (TGFb) and bone morphogenetic protein-2 (BMP2) inhibit myogenin transcriptional activity. Predomitly residing within the nucleus, the subcellular localization of myogenin has been shown to be dependent on differentiation status and cell density. For instance, trafficking of myogenin between the nucleus and cytoplasm has been reported during skeletal muscle differentiation to mediate transcription control. Applications Reported: This F5D antibody has been reported for use in immunoblotting (WB) and immunocytochemistry. Applications Tested: This F5D antibody has been tested by western blot and immunocytochemistry of the C2C12 cell line. This can be used at less than or equal to 1-5 μg/mL for both applications. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC.

Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myogenin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone F5D
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MYOG
Gene Accession No. P12979, P15173, P20428
Gene Alias bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4)
Gene Symbols MYOG
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17928, 29148, 4656, 490224
Target Species Human, Mouse, Rat, Canine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.