Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myoglobin Mouse anti-Human, Rat, Clone: 1A4F7, Proteintech
SDP

Catalog No. 501736638 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6638 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6638 Supplier Proteintech Group Inc Supplier No. 662051IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody well recognized endogenous myoglobin in muscle lysates.

Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.

Specifications

Antigen Myoglobin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A4F7
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MB
Gene Accession No. P02144, Q9QZ76
Gene Alias MB, MYO, myoglobin
Gene Symbols Mb
Host Species Mouse
Immunogen Myoglobin Fusion Protein Ag8979
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4151, 59108
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.