Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Myosin-2 Heavy Chain Peptide, Synthetic, >98% Purity, 1 mg

Catalog No. 502676953
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-267-6953 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-267-6953 Supplier Abcam Supplier No. AB2048581MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic peptide with >98% purity, suitable for functional studies and HPLC.

  • Purity: >98%
  • Form: Lyophilized
  • Applications: FuncS, HPLC
  • Synthetic: Yes

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Myosin-2 Heavy Chain
Content And Storage Store at -20°C.
Form Lyophilized
Label or Dye Unconjugated
Shipping Condition Blue Ice
Substrate Properties Peptide-based Substrate
Quantity 1 mg
For Use With (Application) FuncS, HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.