Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin 7 (MYH7) Mouse anti-Mouse, Human, Clone: BA-D5, MilliporeSigma™

Catalog No. MABT83825UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT838-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT838-25UG Supplier MilliporeSigma Supplier No. MABT83825UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Myosin 7 (MYH7), clone BA-D5 is a mouse monoclonal antibody that detects Myosin-7 and has been tested for use in Immunofluorescence, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin 7 (MYH7)
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone BA-D5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Myh7
Host Species Mouse
Immunogen Purified myosin from bovine atrium.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_777152
Test Specificity Clone BA-D5 detects Myosin-7 in multiple mammalian species. It detects both alpha- and beta-slow myosin heavy chain.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.