Learn More
Myosin Heavy Chain Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Myosin Heavy Chain |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MF20 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative |
| Gene Alias | DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc |
| Gene Symbols | MYH14 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.