Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin 4 Monoclonal Antibody (MF20), eBioscience™, Invitrogen™

Catalog No. p-7069516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2707 25 μg
50-112-2708 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2707 Supplier Invitrogen™ Supplier No. 14650380
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This MF20 monoclonal antibody recognizes the heavy chain of myosin II, specificially the light meromyosin portion, in cardiac and skeletal muscle of vertebrates. Myosin II is composed of two heavy chains and four light chains. The 220-kDa myosin heavy chain exists as four different isoforms due to alternative splicing. Myosins interact with actin and hydrolyze ATP to function in muscle contraction, cytokinesis, and phagocytosis. The MF20 has been shown to react to myosin from a variety of mammalian, avian and amphibian species, including rat, mouse, human, chicken, zebrafish, and dog. Applications Reported: This MF20 antibody has been reported for use in western blotting, and immunocytochemistry. Applications Tested: This MF20 antibody has been tested by immunocytochemistry and western blot and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin 4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MF20
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene MYH4
Gene Accession No. Q076A5, Q28641, Q29RW1, Q5SX39, Q9Y623
Gene Alias 2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product
Gene Symbols MYH4
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009548, 100125991, 100726456, 17884, 360543, 408020, 4622, 468396, 479502
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Feline, Chicken, Guinea Pig, Chimpanzee
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.