Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin Heavy Chain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115216 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115216 Supplier Invitrogen™ Supplier No. PA5115216
  • $592.00
    $490.65 / Each of 1

    Save $101.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Fast Myosin Skeletal Heavy chain.

The function of Mhc is muscle contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin Heavy Chain
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MYH1
Gene Accession No. P02563, P12882, P13533, Q02566, Q5SX40
Gene Alias A530084A17Rik; A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; beta-myosin heavy chain; Bsh; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CG17927; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; chr2L:16765388..16765564; chr2L:16781880..16782040; CMD1EE; CMH14; DFNA4; DFNA4A; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; epididymis luminal protein 71; FP17425; heavy chain; HEL71; ifm(2)2; ifm(2)RU1; ifm(2)RU2; IId; IId/x; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; KIAA2034; l(2)36Ae; l(2)k10423; l(2)M66; MdMs; mhc; MHC16; MHC2X/D; MHC-2X/D; Mhc36B; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; mMHC; MRP; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; MYH1; MYH14; MYH14 variant protein; MYH17; MYH6; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhca; Myhc-a; MyHC-alpha; MyHC-IId/x; MYHC-IIX; myHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; Myo; Myo6; myosin; myosin HC; myosin heavy; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 14; Myosin heavy chain 2x; myosin heavy chain 6; Myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; Myosin heavy chain, muscle; Myosin heavy chain, non-muscle IIc; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, skeletal muscle, adult 1; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin II heavy chain; myosin rod protein; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 14, non-muscle; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 14; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-1; Myosin-14; myosin-6; myosin-heavy chain; NMHC II-C; NMHC-II-C; non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; Not-upheld; Nup; PNMHH; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; Sht; sMHC; SSS3; Stp; stuckup; type 2X myosin heavy chain
Gene Symbols MYH1, Myh6
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYH1(Accession P12882), corresponding to amino acid residues V608-N658.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17879, 17888, 287408, 29556, 4619, 4624
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.