Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin Skeletal Muscle Monoclonal Antibody (MYSN02 (MY-32))
GREENER_CHOICE

Catalog No. PIMA511748 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511748 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511748 Supplier Invitrogen™ Supplier No. MA511748
  • $1,135.00
    $874.65 / Each of 1

    Save $260.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA5-11748 targets Myosin Skeletal Muscle in IHC (P) applications and shows reactivity with Human samples. The MA5-11748 immunogen is rabbit muscle myosin.

Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP. Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains. Myosin heavy chains are encoded by a multigene family. In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated, smooth, and nonmuscle cells. These isoforms show expression that is spatially and temporally regulated during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin Skeletal Muscle
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MYSN02 (MY-32)
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with ≤0.1% sodium azide
Gene Myh1
Gene Accession No. P12882, Q9UKX2, Q9Y623
Gene Alias 2A myosin heavy chain; 2B myosin heavy chain; A530084A17Rik; AI506973; epididymis luminal protein 71; fast 2a myosin heavy chain; fast myosin heavy chain; fast myosin heavy chain 2X; HEL71; IBM3; IId; IId/x; inclusion body myopathy 3, autosomal dominant; MdMs; MGC133384; MHC; MHC2A; MHC2B; MHC2X/D; MHC-2X/D; Minimsc; Minmus; MM; MYH1; MYH2; MYH2A; MYH2B; MYH-2B; Myh4; MYHa; MYHas8; MYHC; MyHC-2A; MyHC-2b; MYHC-2d; MyHC-2x; MyHC-2X/D; MyHC-IIA; MyHC-IIb; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; MYHSA2; Myhsf; Myhs-f; Myhsf1; Myhs-f1; Myhsf2; Myhs-f2; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 2; myosin heavy chain 2a; Myosin heavy chain 2b; myosin heavy chain 2d; myosin heavy chain 2X; Myosin heavy chain 4; myosin heavy chain IIa; Myosin heavy chain IIb; myosin heavy chain IIx/d; myosin heavy chain isoform 2b; myosin heavy chain isoform fast type; myosin heavy chain type IIb; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin, heavy polypeptide 4, skeletal muscle; myosin-1; Myosin-2; myosin-4; MYPOP; skeletal muscle myosin heavy chain II-a; type 2X myosin heavy chain; type IIA myosin heavy chain; unnamed protein product
Gene Symbols Myh1, MYH2, MYH4
Host Species Mouse
Immunogen Rabbit muscle myosin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4619, 4620, 4622
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.