Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ MyPIPETMANTM Volume Display Windows

Catalog No. FP070102G Shop All Gilson Products
Change view
Click to view available options
Color:
Blue
Green
Navy
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
FP070102G White
FP070104G Red
FP070105G Yellow
FP070106G Green
FP070103G Blue
FP070101G Navy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. FP070102G Supplier Gilson™ Supplier No. FP070102
Add to Cart
Add to Cart
This item is not returnable. View return policy

For easier identification and convenience, this colored window provides extra pipette customization.

  • Six available colors
  • Easily replace the colored window at your convenience

Specifications

Autoclavable Autoclavable
Calibrated No
Material Polystyrene, Polypropylene
Sterility Non-sterile
Graduated No
Quantity 1
Color White
Type Display Window
Disposable Reusable
For Use With (Equipment) MyPIPETMAN
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.