Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myrcene, Analytical Standard, ≥90.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018570 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8570 500 mg
11-102-0211 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8570 Supplier MilliporeSigma Supelco Supplier No. 64643500MGF
Add to Cart
Add to Cart

Myrcene is an acyclic monoterpene having antimutagenic properties, which can be found in the essential oils of many useful plants like lemongrass, hop, bay, verbena and also others.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 123-35-3
Vapor Pressure ˜7 mmHg (20°C)
Vapor Density 4.7 (vs. air)
Density 0.791 g/mL (at 25°C)
Physical Form Neat
Linear Formula H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
MDL Number MFCD00008908
Boiling Point 167°C
Synonym beta-Myrcene; 7-Methyl-3-methylene-1,6-octadiene
Quantity 500 mg
Formula Weight 136.23
Percent Purity ≥90.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.