Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myricitrin 98.0+%, TCI America™
SDP

Catalog No. M236110MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M236110MG 10 mg
M236150MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M236110MG Supplier TCI America Supplier No. M236110MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17912-87-7
Molecular Formula C21H20O12
Molecular Weight (g/mol) 464.379
MDL Number MFCD00016930
InChI Key DCYOADKBABEMIQ-OWMUPTOHSA-N
PubChem CID 5281673
ChEBI CHEBI:70082
IUPAC Name 5,7-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
SMILES CC1C(C(C(C(O1)OC2=C(OC3=CC(=CC(=C3C2=O)O)O)C4=CC(=C(C(=C4)O)O)O)O)O)O

Specifications

Melting Point 197°C
Quantity 10 mg
Synonym 3,3′C,4′C,5,5′C,7-Hexahydroxyflavone 3-O-Rhamnoside
Formula Weight 464.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Myricitrin
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.