Learn More
Enzo Life Sciences Myriocin (5 mg) CAS: 35891-70-4
Supplier: Enzo Life Sciences BMLSL2260005
Palmitoyltransferase inhibitor. Potent immunosuppressant ~10 to 100-fold more potent than cyclosporin A (Prod. No. BML-A195). Blocks sphingolipid biosynthesis by inhibiting serine palmitoyltransferase (Ki=0.28nM), therefore useful as a tool to deplete cells of sphingolipids. Induces apoptosis by depletion of cellular sphingolipids. Inhibits proliferation of an IL-2-dependent mouse cytotoxic cell line CTLL-1 (IC50=15nM). Synonyms: ISP-1, Thermozymocidin, 2S,3R,4R,6E-2-Amino-3,4-dihydroxy-2-hydroxymethyl-14-oxo-6-eicosenoic acid. Formula: C21H39NO6. MW: 401.5.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.