Learn More
Cayman Chemical ANTIBODY BQ-788 500ug
SDP
Supplier:  Cayman Chemical 31156500 UG
Encompass_Preferred
An inhibitor that blocks the integrin-ligand interaction between LFA-1 and ICAM-1 (IC50 35 nM) inhibits adhesion of JY-8 cells to ICAM-1-coated microtiter plates (IC50 44 nM)
Catalog No. 50-299-0377
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.