Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Myrtenol, ≥98.5% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018655 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8655 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8655 Supplier MilliporeSigma Supelco Supplier No. 701581ML

(−)-Myrtenol is a naturally occurring monoterpene alcohol, belonging to the class of volatile organic compounds. It is widely used in fragrance, cosmetic products, cleaning agents and flavoring agents in a variety of food products.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 19894-97-4
Density 0.981 g/mL (at 20°C)
Flash Point 68°C
Physical Form Neat
Linear Formula C10H16 O
MDL Number MFCD00001341
Boiling Point 220°C to 222°C
Synonym (1 R)-2-Pinen-10-ol; (1 R)-6,6-Dimethylbicyclo[3.1.1 ]hept-2-ene-2-methanol
Type Analytical Standard
Quantity 1 mL
Formula Weight 152.23
Percent Purity ≥98.5% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.