Learn More
Eisco™ Mystery Density Set With Stand, 12 Bars
SCI_ED
Catalog No. S12926 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
S12926 12 Pk.
1 options
This item is not returnable. View return policy
Catalog No. S12926 Supplier Eisco™ Supplier No. PH0113MDS
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Mystery Density Set With Stand is an experiment of intrigue as students will need to use the scientific method and understanding of density to determine the material of the rods in this set.
  • Students will have to determine what the unknown substance is by taking careful volume and mass measurements in order to calculate density and determine the mystery material.
  • To determine the density of the material the student plots mass versus volume and the slope therefore is the density (mass per unit volume).
  • Students will need to do all measurements themselves so a balance and ruler are required.
  • Base is made from a durable wood coated for years of use.
  • Machined from top quality raw materials to produce accurate results and durability of decades of use.
Specifications
Quantity 12 Pk.
Product Type Cylinder Set
Includes Stand