Learn More
Medchemexpress LLC MZ-101 (GYS1-IN-2) | 2839908-40-4 | 99.0% | 449.52 | C25H28FN5O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15764610MG
Encompass_Preferred
MZ-101 (GYS1-IN-2) is an orally active, potent, and selective small-molecule inhibitor of glycogen synthase 1 (GYS1) with an IC50 of 0.041 μM. It reduces glycogen concentrations in cells and in mice and is used to study GYS1-mediated Pompe disease and other glycogen storage disorders.
  • Potent, selective GYS1 inhibition (IC50 = 0.041 μM).
  • Reduces glycogen concentrations in cells and animal models.
  • Orally active small molecule suitable for in vivo studies.
  • High purity (98.98%) and solid physical form for storage stability.
  • Available in small solid quantities and DMSO solutions for flexible dosing.
  • Storage: powder stable at -20 °C (up to three years) and 4 °C (up to two years).
Catalog No. 50-002-87993