Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals™

Catalog No. MP215125225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215125225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215125225 Supplier MP Biomedicals, Inc Supplier No. 0215125225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as an ion pairing reagent for HPLC and as an anionic surfactant. N-1-Hexanesulfonic Acid Sodium Salt Monohydrate, MP Biomedicals is a low molecular weight alkylsulfonate. The anionic sulfonate counterion permits the separation and resolution of posit

Because of its low molecular weight, Sodium hexanesulfonate will not form micelles in solution and therefore has no CMC value. This product is used for ion-pair chromatography. It is also utilized in the analysis of small organic compounds, pharmaceutical products and metabolites by such techniques as HPLC.

TRUSTED_SUSTAINABILITY

Specifications

Form White Flakes
Quantity 25 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.