Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-(1-Naphthyl)ethylenediamine Dihydrochloride, Coupling Agent, Solid, 10 g

Catalog No. 502704657
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4657 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4657 Supplier Abcam Supplier No. AB14639510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coupling agent used for spectrophotometric determination and quantitative analysis of nitrate and nitrite in water samples.

  • Molecular Weight: 259.18 g/mol
  • Form: Solid
  • Applications: Coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols, and sulfonamides; quantitative analysis of nitrate and nitrite in water samples by colorimetry
  • Constituents: 100% N-2-Aminoethyl-1-naphthylamine dihydrochloride
  • Storage: Store at ambient temperature
  • CAS Number: 1465-25-4

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Aromatic Amines, Aminophenols, Phenylenediamines, Dinitroanilines, Chloroanilines, Thiols, Sulfonamides, Nitrate, Nitrite
Shipping Condition Ambient
Form Solid
Content And Storage Store at ambient temperature.
Detection Method Colorimetric
Sample Type Water
Quantity 10 g
For Use With (Application) Spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides, quantitative analysis of nitrate and nitrite in water samples by colorimetry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.