Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride for determination of sulfonamide and nitrite, ACS reagent, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111013139 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3139 25 g
11-101-3140 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3139 Supplier MilliporeSigma Supelco Supplier No. 3346125G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00012556
Synonym 2-(1-Naphthylamino)ethylamine dihydrochloride

Specifications

Melting Point 194°C to 198°C
CAS 1465-25-4
Linear Formula C10H7 NHCH2 CH2 NH2 · 2 HCl
Quantity 25 g
Formula Weight 259.17
Percent Purity ≥98% (calc with ref. to anhyd. subst.); ≥98%
Grade ACS Reagent
RTECSNumber KV5330000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.