Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane 90.0+%, TCI America™
SDP

Catalog No. B4062100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4062100MG 100 mg
B406225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4062100MG Supplier TCI America Supplier No. B4062100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1263166-93-3
Molecular Formula C17H28N2O4
Molecular Weight (g/mol) 324.421
MDL Number MFCD19705418
InChI Key YZGOWXGENSKDSE-XYPWUTKMSA-N
Synonym BCN-POE3-NH2, BCN-amine
PubChem CID 75412389
IUPAC Name [(1S,8R)-9-bicyclo[6.1.0]non-4-ynyl]methyl N-[2-[2-(2-aminoethoxy)ethoxy]ethyl]carbamate
SMILES C1CC2C(C2COC(=O)NCCOCCOCCN)CCC#C1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 324.42
Percent Purity ≥90.0% (HPLC)
Physical Form Liquid
Chemical Name or Material N-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethyloxycarbonyl-1,8-diamino-3,6-dioxaoctane
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.