Learn More
Sigma Organic Chemistry n 2 3-trimethY-2-isopro | 250mg | 51115-67-4 | MFCD00130070
Supplier: Sigma Organic Chemistry CDS003481250MG

n 2 3-trimethY-2-isopro , 250mg
About this Item:
CAS #: 51115-67-4
MDL #: MFCD00130070
Molecular Weight: 171.28
UNSPSC Code: 12352200
Chemical Formula: C10H21NO
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.