Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2,4-Dinitrophenyl)-L-serine 98.0+%, TCI America™
SDP

Catalog No. D10371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10371G 1 g
D1037100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10371G Supplier TCI America Supplier No. D10371G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1655-64-7
Molecular Formula C9H9N3O7
Molecular Weight (g/mol) 271.185
MDL Number MFCD00038098
InChI Key SBQZBOCQYMVLTC-ZETCQYMHSA-N
Synonym n-2,4-dinitrophenyl-l-serine, dnp-ser, n-dnp-l-serine, 2s-2-2,4-dinitrophenyl amino-3-hydroxypropanoic acid, 1oau, serine, n-2,4-dinitrophenyl-, dl, dnp-ser-oh, ser-dnf, n-2-4-dnp-l-serine, n-2,4-dnp-l-serine
PubChem CID 96800
ChEBI CHEBI:53084
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NC(CO)C(=O)O

Specifications

Melting Point 177°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 271.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2,4-Dinitrophenyl)-L-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.