Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Chloroethyl)dibenzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C016825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C016825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C016825G Supplier TCI America Supplier No. C016825G
Add to Cart
Add to Cart

Specifications

CAS 55-43-6
Melting Point 190°C
Color White
Molecular Formula C16H19Cl2N
Quantity 25 g
MDL Number MFCD00012518
Synonym dibenamine hydrochloride, n-2-chloroethyl dibenzylamine hydrochloride, sympatholytin, n,n-dibenzyl-2-chloroethanamine hydrochloride, n,n-dibenzyl-2-chloroethylamine hydrochloride, dibenzylchlorethamine hydrochloride, dibenzylchlorethylamine hydrochloride, n,n-dibenzylaminoethyl chloride hydrochloride, 2-chloroethyldibenzylamine hydrochloride, 2-dibenzylaminoethyl chloride hydrochloride
InChI Key LZXCEBPGNFLHEQ-UHFFFAOYSA-N
SMILES [H+].[Cl-].ClCCN(CC1=CC=CC=C1)CC1=CC=CC=C1
IUPAC Name hydrogen dibenzyl(2-chloroethyl)amine chloride
Molecular Weight (g/mol) 296.24
PubChem CID 5925
Formula Weight 296.24
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Chloroethyl)dibenzylamine Hydrochloride
Show More Show Less
RTECSNumber HQ6825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.