Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[2-(Dimethylamino)ethyl]-N-methylformamide 97.0+%, TCI America™
SDP

Catalog No. D372425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D372425G 25 g
D37245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D372425G Supplier TCI America Supplier No. D372425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 105669-53-2
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.191
MDL Number MFCD04039895
InChI Key CSLBJRKWKVBRSQ-UHFFFAOYSA-N
PubChem CID 279482
IUPAC Name N-[2-(dimethylamino)ethyl]-N-methylformamide
SMILES CN(C)CCN(C)C=O

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
Synonym N-Formyl-N,N′,N′-trimethylethylenediamine
Formula Weight 130.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[2-(Dimethylamino)ethyl]-N-methylformamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.