Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Pyridyl)pivalamide 98.0+%, TCI America™
SDP

Catalog No. P238525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P238525G 25 g
P23855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P238525G Supplier TCI America Supplier No. P238525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86847-59-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00460277
InChI Key CGSPVYCZBDFPHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-2-yl-propionamide, 2-pivaloylamino pyridine, 2,2-dimethyl-n-pyridin-2-yl propanamide, n-pyridin-2-yl pivalamide, n-boc-2-aminopyridine, 2,2-dimethyl-n-2-pyridyl propanamide, 2,2-dimethyl-n-2-pyridinyl propanamide, propanamide, 2,2-dimethyl-n-2-pyridinyl, pivaloylamino pyridine, n-o-pyridylpivalamide
PubChem CID 1051519
IUPAC Name 2,2-dimethyl-N-pyridin-2-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=CC=N1

Specifications

Melting Point 73°C
Quantity 25 g
Formula Weight 178.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Pyridyl)pivalamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.