Learn More
Abcam N-(3-Hydroxydodecanoyl)-DL-homoserine Lactone, Quorum Sensing Agent, >97% Purity, 25 mg
Description
- Molecular Weight: 299.41 g/mol
- Purity: >97%
- Product Type: Quorum sensing agent
- Family: Member of N-acyl-homoserine lactone (AHL) family, which regulates gene expression in gram-negative bacteria and is involved in bacterial quorum sensing processes
- Storage Temperature: Store at Ambient
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | N-(3-Hydroxydodecanoyl)-DL-homoserine Lactone |
| Form | Solid |
| Buffer | 100% N-(3-Hydroxydodecanoyl)-DL-homoserine lactone |
| Content And Storage | Store at ambient temperature. |
| Quantity | 25 mg |
| For Use With (Application) | Bacterial quorum sensing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.