Learn More
Abcam N-(3-Hydroxydodecanoyl)-DL-homoserine Lactone, Quorum Sensing Agent, >97% Purity, 250 mg
Description
- Molecular weight: 299.41 g/mol
- Purity: >97%
- Quorum sensing agent: Member of N-acyl-homoserine lactone (AHL) family
- Functional use: Regulates gene expression in gram-negative bacteria and is involved in bacterial quorum sensing processes
- Storage: Store at ambient
- Shipping: Ambient - Can ship with ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N-(3-Hydroxydodecanoyl)-DL-homoserine lactone |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% N-(3-Hydroxydodecanoyl)-DL-homoserine lactone |
| Format | Vial |
| Formulation | 100% N-(3-Hydroxydodecanoyl)-DL-homoserine lactone |
| Quantity | 250 mg |
| For Use With (Application) | Bacterial quorum sensing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.