Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Hydroxypropyl)ethylenediamine 97.0+%, TCI America™
SDP

Catalog No. H039125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H039125G 25 g
H03915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H039125G Supplier TCI America Supplier No. H039125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56344-32-2
Molecular Formula C5H14N2O
Molecular Weight (g/mol) 118.18
MDL Number MFCD00059848
InChI Key KTLIZDDPOZZHCD-UHFFFAOYSA-N
Synonym N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine
PubChem CID 10973740
IUPAC Name 3-(2-aminoethylamino)propan-1-ol
SMILES C(CNCCN)CO

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 149°C
Quantity 25 g
UN Number 3259
Formula Weight 118.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N-(3-Hydroxypropyl)ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.