Learn More
Abcam N-(3-Oxododecanoyl)-L-homoserine Lactone, >98% Purity, Powder, 10 mg
Description
- Molecular Weight: 297.39 g/mol
- Purity: >98%
- Form: Powder
- Storage: Store at Ambient
- Shipping: Ambient - Can Ship with Ice
- Description: Autoinducer involved in quorum sensing by Pseudomonas aeruginosa. Member of the N-acyl-homoserine lactone (AHL) family that regulates gene expression in gram-negative bacteria and is involved in the processes of bacterial quorum sensing.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | N-(3-Oxododecanoyl)-L-homoserine Lactone |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% N-(3-Oxo-dodecanoyl) homoserine lactone |
| Content And Storage | Store at ambient temperature. |
| Form | Powder |
| Formulation | 100% N-(3-Oxo-dodecanoyl) homoserine lactone |
| Quantity | 10 mg |
| For Use With (Application) | Bacterial quorum sensing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.