Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-(3-Oxododecanoyl)-L-homoserine Lactone, >98% Purity, Powder, 100 mg

Catalog No. 502705152
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5152 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5152 Supplier Abcam Supplier No. AB145509100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity autoinducer involved in Pseudomonas aeruginosa quorum sensing.

  • Molecular Weight: 297.39 g/mol
  • Purity: >98%
  • Form: Powder
  • Storage: Ambient
  • Application: Autoinducer involved in quorum sensing by Pseudomonas aeruginosa
  • Description: Member of the N-acyl-homoserine lactone (AHL) family that regulates gene expression in gram-negative bacteria and is involved in bacterial quorum sensing processes

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule N-(3-Oxododecanoyl)-L-homoserine Lactone
Shipping Condition Ambient
Buffer 100% N-(3-Oxo-dodecanoyl) homoserine lactone
Content And Storage Store at ambient temperature.
Form Powder
Formulation 100% N-(3-Oxo-dodecanoyl) homoserine lactone
Quantity 100 mg
For Use With (Application) Bacterial quorum sensing, gene expression regulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.