Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-(3-Oxododecanoyl)-L-homoserine Lactone, >98% Purity, Powder, 20 mg

Catalog No. 502705150
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5150 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5150 Supplier Abcam Supplier No. AB14550920MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Autoinducer involved in quorum sensing in Pseudomonas aeruginosa.

  • Molecular weight: 297.39 g/mol
  • Purity: >98%
  • Physical form: Powder
  • Components: 100% N-(3-Oxo-dodecanoyl) homoserine lactone
  • Function: Autoinducer involved in quorum sensing by Pseudomonas aeruginosa and gene expression regulation in gram-negative bacteria
  • Citations: 1

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule N-(3-Oxododecanoyl)-L-homoserine Lactone
Content And Storage Store at ambient temperature.
Form Powder
Formulation 100% N-(3-Oxo-dodecanoyl) homoserine lactone
Quantity 20 mg
For Use With (Application) Bacterial quorum sensing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.