Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobenzoyl)-L-glutamic Acid 97.0+%, TCI America™
SDP

Catalog No. A04421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04421G 1 g
A04425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04421G Supplier TCI America Supplier No. A04421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4271-30-1
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.253
MDL Number MFCD00042821
InChI Key GADGMZDHLQLZRI-VIFPVBQESA-N
PubChem CID 196473
ChEBI CHEBI:61114
IUPAC Name (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)N

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 266.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic Acid
EINECSNumber (9)-1580
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.