Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobutyl)-2-naphthalenesulfonamide hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111015754 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5754 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5754 Supplier MilliporeSigma Supelco Supplier No. A31681MG
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 89108-46-3
Physical Form Powder
Linear Formula C14H18N2O2 S · HCl
MDL Number MFCD00058052
Synonym W-12
Solubility Soluble in water
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 mg
Formula Weight 314.83
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.