Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobutyl)-2-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A240825MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A240825MG 25 mg
A2408500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A240825MG Supplier TCI America Supplier No. A240825MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89108-46-3
Molecular Formula C14H19ClN2O2S
Molecular Weight (g/mol) 314.828
MDL Number MFCD00058052
InChI Key WYFVKUWCEVMLOL-UHFFFAOYSA-N
Synonym W-12 Hydrochloride
PubChem CID 9839864
IUPAC Name N-(4-aminobutyl)naphthalene-2-sulfonamide;hydrochloride
SMILES C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)NCCCCN.Cl

Specifications

Melting Point 183°C
Color White-Yellow
Quantity 25 mg
Formula Weight 314.83
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobutyl)-2-naphthalenesulfonamide Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.