Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A241625MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A241625MG 25 mg
A2416500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A241625MG Supplier TCI America Supplier No. A241625MG

Chemical Identifiers

CAS 88519-57-7
Molecular Formula C14H18Cl2N2O2S
Molecular Weight (g/mol) 349.27
MDL Number MFCD00058030
InChI Key QKAALLVQBOLELJ-UHFFFAOYSA-N
Synonym w-13 hydrochloride, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamide hydrochloride, w-13, hydrochloride, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hydrochloride, 2-naphthalenesulfonamide,n-4-aminobutyl-5-chloro-, hydrochloride 1:1, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide, hcl, acmc-209qtx, n-4-aminobutyl-5-chloro-2-naphthalenesulfonamide hcl, n-4-aminobutyl-5-chloronaphthalene-2-sulfonamidehydrochloride, n-4-aminobutyl-5-chloro-2-naphthalene sulfonamide hydro chloride
PubChem CID 16760706
ChEBI CHEBI:75367
IUPAC Name N-(4-aminobutyl)-5-chloronaphthalene-2-sulfonamide;hydrochloride
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)NCCCCN)C(=C1)Cl.Cl

Specifications

Melting Point 237°C
Quantity 25 mg
Formula Weight 349.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.