Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Bromophenyl)phthalimide 98.0+%, TCI America™
SDP

Catalog No. B270725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B270725G 25 g
B27075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B270725G Supplier TCI America Supplier No. B270725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40101-31-3
Molecular Formula C14H8BrNO2
Molecular Weight (g/mol) 302.13
MDL Number MFCD00047301
InChI Key DNQCPYWSXMATIN-UHFFFAOYSA-N
PubChem CID 101305
IUPAC Name 2-(4-bromophenyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES BrC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 206°C
Color White
Quantity 25 g
Formula Weight 302.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Bromophenyl)phthalimide
RTECSNumber TI4025000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.