Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 98.0+%, TCI America™
SDP

Catalog No. C33151G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33151G 1 g
C33155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33151G Supplier TCI America Supplier No. C33151G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 848133-76-6
Molecular Formula C14H12ClN3O2
Molecular Weight (g/mol) 289.72
MDL Number MFCD09263704
InChI Key XDXGFTCQRAQEEG-UHFFFAOYSA-N
PubChem CID 11358234
IUPAC Name N-(4-chloro-3-cyano-7-ethoxyquinolin-6-yl)acetamide
SMILES CCOC1=C(NC(C)=O)C=C2C(Cl)=C(C=NC2=C1)C#N

Specifications

Melting Point 250°C
Quantity 1 g
Formula Weight 289.72
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.