Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Nitrophenyl)-3-oxobutyramide 98.0+%, TCI America™
SDP

Catalog No. N09901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09901G 1 g
N0990200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09901G Supplier TCI America Supplier No. N09901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4835-39-6
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.2
MDL Number MFCD00452522
InChI Key KCXJQNDNGLRYBN-UHFFFAOYSA-N
Synonym 4′C-Nitroacetoacetanilide
PubChem CID 78547
IUPAC Name N-(4-nitrophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 122°C
Color Yellow
Quantity 1 g
Formula Weight 222.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Nitrophenyl)-3-oxobutyramide
EINECSNumber (3)-0393
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.