Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(5-Bromopentyl)phthalimide 98.0+%, TCI America™
SDP

Catalog No. B498625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B498625G 25 g
B49865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B498625G Supplier TCI America Supplier No. B498625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 954-81-4
Molecular Formula C13H14BrNO2
Molecular Weight (g/mol) 296.164
MDL Number MFCD00060522
InChI Key QKVHAKICMNABGB-UHFFFAOYSA-N
Synonym n-5-bromopentyl phthalimide, 2-5-bromopentyl isoindole-1,3-dione, n-5-bromopentyl-phthalimide, 2-5-bromopentyl isoindoline-1,3-dione, 2-5-bromo-pentyl-isoindole-1,3-dione, 2-5-bromopentyl-1h-isoindole-1,3 2h-dione, 2-5-bromopentyl-2,3-dihydro-1h-isoindole-1,3-dione, 5-phthalimidopentyl bromide, 1-bromo-5-phthalimidopentane
PubChem CID 136770
IUPAC Name 2-(5-bromopentyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCBr

Specifications

Melting Point 62°C
Quantity 25 g
Formula Weight 296.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(5-Bromopentyl)phthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.