Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine 98.0+%, TCI America™
SDP

Catalog No. F10131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10131G 1 g
F1013200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10131G Supplier TCI America Supplier No. F10131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 159611-02-6
Molecular Formula C22H19NO5
Molecular Weight (g/mol) 377.396
MDL Number MFCD01311747
InChI Key AJXDCHXGNUFBRC-FQEVSTJZSA-N
Synonym fmoc-l-2-furylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-ala-oh, fmoc-3-2-furyl-l-alanine, fmoc-l-3-2-furyl-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, rarechem bk pt 0223, s-n-fmoc-furylalanine, fmoc-3-ala 2-furyl-oh
PubChem CID 2734451
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 1 g
Formula Weight 377.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-L-alanine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.