Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. F091125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F091125G 25 g
F09115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F091125G Supplier TCI America Supplier No. F091125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 116821-47-7
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
MDL Number MFCD00144889
InChI Key ACUIFAAXWDLLTR-UHFFFAOYSA-N
Synonym fmoc-gaba-oh, fmoc-gamma-abu-oh, fmoc-gaba, n-fmoc-dl-4-amino-butyric acid, 4-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, 4-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-fmoc-4-aminobutyric acid, 4-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-gamma-fmoc-gamma-aminobutyric acid, 4-9h-fluoren-9-yl methoxy carbonylamino butanoic acid
PubChem CID 2756086
IUPAC Name 4-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCCC(=O)O

Specifications

Melting Point 174°C
Color White
Quantity 25 g
Formula Weight 325.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.