Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. F045525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F045525G 25 g
F04551G 1 g
F04555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F045525G Supplier TCI America Supplier No. F045525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73731-37-0
Molecular Formula C19H21NO6
Molecular Weight (g/mol) 359.38
MDL Number MFCD00792901
InChI Key CGNQPFAECJFQNV-NRNQBQMASA-N
Synonym fmoc-thr-oh, fmoc-l-threonine, fmoc-l-thr-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-hydroxy-butanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid, n-9-fluorenylmethoxycarbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid, fmoc-thr-oh monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine monohydrate
PubChem CID 6992530
IUPAC Name (2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid hydrate
SMILES O.C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 341.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine Monohydrate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.